Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-(Allyloxycarbonyloxy)succinimide 97.0+%, TCI America™

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Color | White |
Quantity | 25 g |
Formula Weight | 199.16 |
Percent Purity | ≥97.0% (GC) |
Physical Form | Crystalline Lumps |
Chemical Name or Material | N-(Allyloxycarbonyloxy)succinimide |
Safety and Handling
TSCA | No |
Recommended Storage | Refrigerator |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction