Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Allylphthalimide 98.0+%, TCI America™
SDP

Catalog No. A26961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26961G 1 g
A2696200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26961G Supplier TCI America Supplier No. A26961G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5428-09-1
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.20
MDL Number MFCD00158662
InChI Key MHHGQWMCVNQHLG-UHFFFAOYSA-N
Synonym 2-allylisoindoline-1,3-dione, n-allylphthalimide, n-2-propenyl phthalimide, 2-allyl-1h-isoindole-1,3 2h-dione, 2-prop-2-en-1-yl-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 2-2-propen-1-yl, 2-prop-2-en-1-yl-2,3-dihydro-1h-isoindole-1,3-dione, n-allylphthalimid, n-allyl-phthalimide, 2-allylisoindole-1,3-dione
PubChem CID 224401
IUPAC Name 2-prop-2-enylisoindole-1,3-dione
SMILES C=CCN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 70°C
Color White-Yellow
Boiling Point 295°C
Quantity 1 g
Formula Weight 187.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Allylphthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.