Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, 98%

Catalog No. ac439341000
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle

CAS: 95713-52-3 | C9H9FN4O5 | 272.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95713-52-3
Molecular Formula C9H9FN4O5
Molecular Weight (g/mol) 272.19
MDL Number MFCD00042049
InChI Key NEPLBHLFDJOJGP-BYPYZUCNSA-N
Synonym marfey's reagent, s-2-5-fluoro-2,4-dinitrophenyl amino propanamide, 2s-2-5-fluoro-2,4-dinitrophenyl amino propanamide, n-2,4-dinitro-5-fluorophenyl-l-alaninamide, 2s-2-5-fluoro-2,4-dinitroanilino propanamide, propanamide, 2-5-fluoro-2,4-dinitrophenyl amino-, 2s, nalpha-2,4-dinitro-5-fluorophenyl-l-alaninamide, n-alpha-2,4-dinitro-5-fluorophenyl-l-alaninamide, fdaa, 1-fd-aa
PubChem CID 5486955
IUPAC Name (2S)-2-(5-fluoro-2,4-dinitroanilino)propanamide
SMILES C[C@H](NC1=CC(F)=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(N)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 227.7°C to 229.4°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Specific Rotation +54.0° to +58.0° (20°C, 589nm) (c=1, acetone)
Formula Weight 272.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-α-(2, 4-Dinitro-5-fluorophenyl)-L-alaninamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.