Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[N-[alpha-[2-(Dibutylglycinamido)phenyl]benzylidene]glycinato]nickel 95.0+%, TCI America™
SDP

Catalog No. D3543100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3543100MG 100 mg
D35431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3543100MG Supplier TCI America Supplier No. D3543100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 847654-16-4
Molecular Formula C25H31N3NiO3
Molecular Weight (g/mol) 480.23
MDL Number MFCD10566890
InChI Key KZZGXIZCIUYKSB-UHFFFAOYSA-L
PubChem CID 53384304
IUPAC Name nickel(2+) 2-{[(2-{[2-(dibutylamino)-1-oxidoethylidene]amino}phenyl)(phenyl)methylidene]amino}acetate
SMILES [Ni++].CCCCN(CCCC)CC([O-])=NC1=CC=CC=C1C(=NCC([O-])=O)C1=CC=CC=C1

Specifications

Quantity 100 mg
Formula Weight 480.23
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [N-[alpha-[2-(Dibutylglycinamido)phenyl]benzylidene]glycinato]nickel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.