Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Amyl acetate 99%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
628-63-7
C7H14O2
130.19
MFCD00009500
PGMYKACGEOXYJE-UHFFFAOYSA-N
amyl acetate,n-amyl acetate,acetic acid, pentyl ester,n-pentyl acetate,amyl acetic ester,1-pentyl acetate,n-pentyl ethanoate,birnenoel,pent-acetate,amyl acetic ether
12348
CHEBI:87362
pentyl acetate
CCCCCOC(=O)C
628-63-7
C7H14O2
130.19
MFCD00009500
PGMYKACGEOXYJE-UHFFFAOYSA-N
amyl acetate,n-amyl acetate,acetic acid, pentyl ester,n-pentyl acetate,amyl acetic ester,1-pentyl acetate,n-pentyl ethanoate,birnenoel,pent-acetate,amyl acetic ether
12348
CHEBI:87362
pentyl acetate
CCCCCOC(=O)C
Infrared Spectrum Authentic
Refractive Index 1.4010 to 1.4030
Specific Gravity 0.87
Melting Point -70.8°C
Flash Point 24°C
Packaging Glass bottle
Density 0.8700g/mL
Quantity 250mL
Color Colorless
Boiling Point 149.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3CO2(CH2)4CH3
Beilstein 02, 131
Solubility Information Solubility in water: 10g/L (20c). Other solubilities: miscible with alcohol and ether
Formula Weight 130.19
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material n-Amyl acetate
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning