Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-N-ethyl-3-methylaniline, 98%, Thermo Scientific™

Catalog No. p-7056969
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-94-8
Molecular Formula C16H19N
Molecular Weight (g/mol) 225.34
MDL Number MFCD00025950
InChI Key VIACAIAQHPLINF-UHFFFAOYSA-N
Synonym n-benzyl-n-ethyl-m-toluidine, n-ethyl-n-benzyl-m-toluidine, ethylbenzyltoluidine, benzenemethanamine, n-ethyl-n-3-methylphenyl, 3-n-benzyl-n-ethylamino toluene, n-ethyl-n-benzyl-meta-toluidine, 3-n-benzyl-n-ethylamino methylbenzene, m-toluidine, n-benzyl-n-ethyl, n-benzyl-n-ethyl-meta-toluidine, n-ethyl-n-m-tolyl benzylamine
PubChem CID 67076
IUPAC Name N-benzyl-N-ethyl-3-methylaniline
SMILES CCN(CC1=CC=CC=C1)C1=CC=CC(C)=C1

Specifications

Density 0.77 g/mL
Boiling Point 161°C to 162°C
Flash Point 77°C (171°F)
Quantity 1 g
UN Number UN2753
Beilstein 5336067
Formula Weight 225.33
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Benzyl-N-ethyl-3-methylaniline
Hazard Category H227-H301+H311-H330
Hazard Statement GHS H Statement
H301-H311-H331-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: N-ETHYLBENZYLTOLUIDINES LIQUID
EINECSNumber 204-359-6
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.