Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Benzyl-N-methylethanolamine

Catalog No. 366412100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-641-2100G 100 g
36-641-2500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-641-2100G Supplier Sigma Aldrich Supplier No. 366412100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N-benzyl-n-methylethanolamine is a tertiary benzylamine
  • Electrochemical alkoxylation of n-benzyl-n-methylethanolamine is reported
  • Rate of proton exchange of n-benzyl-n-methylethanolamine in aqueous hcl has been reported

Specifications

CAS 101-98-4
Density 1.017 g/mL (at 25°C (literature))
Boiling Point 95°C to 105°C (2 mmHg)
Molecular Formula C10H15NO
Refractive Index n20/D 1.529 (literature)
Linear Formula C6H5CH2N(CH3)CH2CH2OH
MDL Number MFCD00002847
Quantity 100 g
Synonym 2-(N-Benzyl-N-methylamino)ethanol
Molecular Weight (g/mol) 165.23
Percent Purity 90%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.