Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-N-tert-butyl-3-nitrobenzamide, 97%, Thermo Scientific™

Catalog No. AAH5932403
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349091-53-8
Molecular Formula C18H20N2O3
Molecular Weight (g/mol) 312.369
MDL Number MFCD02973642
InChI Key NOSAZZBVBDAHIZ-UHFFFAOYSA-N
PubChem CID 3280016
IUPAC Name N-benzyl-N-tert-butyl-3-nitrobenzamide
SMILES CC(C)(C)N(CC1=CC=CC=C1)C(=O)C2=CC(=CC=C2)[N+](=O)[O-]

Specifications

Quantity 1 g
Formula Weight 312.36
Percent Purity 97%
Chemical Name or Material N-Benzyl-N-tert-butyl-3-nitrobenzamide
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.