Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-p-anisidine 98.0+%, TCI America™
SDP

Catalog No. B040925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B040925G 25 g
B04095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B040925G Supplier TCI America Supplier No. B040925G

Chemical Identifiers

CAS 17377-95-6
Molecular Formula C14H15NO
Molecular Weight (g/mol) 213.28
MDL Number MFCD00059298
InChI Key LIJJGMDKVVOEFT-UHFFFAOYSA-N
Synonym n-benzyl-p-anisidine, n-4-methoxyphenyl-n-benzylamine, 4-methoxyphenyl benzylamine, n-benzyl-para-anisidine, acmc-1bvsm, n-benzyl-p-methoxyaniline, cambridge id 5107549, n-benzyl-4-methoxy-aniline, n-4-methoxyphenyl benzylamine, n-4-methoxyphenyl benzyl amine
PubChem CID 519413
IUPAC Name N-benzyl-4-methoxyaniline
SMILES COC1=CC=C(NCC2=CC=CC=C2)C=C1

Specifications

Melting Point 48°C
Color Yellow
Boiling Point 238°C
Quantity 25 g
Formula Weight 213.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzyl-p-anisidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.