Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylethylenediamine 97.0+%, TCI America™
SDP

Catalog No. B110625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B110625ML 25 mL
B11065ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B110625ML Supplier TCI America Supplier No. B110625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9-4-4152
Molecular Formula C9H14N2
Molecular Weight (g/mol) 150.23
MDL Number MFCD00041896
InChI Key ACYBVNYNIZTUIL-UHFFFAOYSA-N
Synonym n-benzylethylenediamine, n1-benzylethane-1,2-diamine, 2-aminoethyl benzyl amine, ethylenediamine, n-benzyl, 1,2-ethanediamine, n-phenylmethyl, n-2-aminoethyl-n-benzylamine, 2-benzylaminoethylamine, n-benzylethane-1,2-diamine, 1,2-ethanediamine, n1-phenylmethyl, 2-aminoethyl benzylamine
PubChem CID 77801
IUPAC Name N'-benzylethane-1,2-diamine
SMILES C1=CC=C(C=C1)CNCCN

Specifications

Color Yellow
Boiling Point 165°C
Quantity 25 mL
UN Number 2735
Formula Weight 150.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Benzylethylenediamine
EINECSNumber (3)-0312&(9)-1312
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.