Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylglycine ethyl ester hydrochloride, 98%, Thermo Scientific™

Catalog No. AC271370250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 6344-42-9
Molecular Formula C11H15NO2·HCl
Molecular Weight (g/mol) 229.71
MDL Number MFCD00144850
InChI Key ZYCCGXGBRHRRLQ-UHFFFAOYSA-N
Synonym ethyl 2-benzylamino acetate hydrochloride, n-benzylglycine ethyl ester hydrochloride, ethyl 2-benzylamino acetate, chloride, n-benzylglycine ethyl ester hcl, ethyl2-benzylamino acetate hydrochloride, benzylamino-acetic acid ethyl ester hydrochloride, ethyl 2-benzylamino acetate hydrochloridyl
PubChem CID 2724714
IUPAC Name ethyl 2-(benzylamino)acetate;hydrochloride
SMILES CCOC(=O)CNCC1=CC=CC=C1.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 96°C to 100°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5CH2NHCH2CO2C2H5·HCl
Quantity 25 g
Formula Weight 229.71
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Benzylglycine ethyl ester hydrochloride, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.