Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylidenemethylamine, 98+%

Catalog No. AAB2516114
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 622-29-7 | C8H9N | 119.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-29-7
Molecular Formula C8H9N
Molecular Weight (g/mol) 119.167
MDL Number MFCD00008294
InChI Key HXTGGPKOEKKUQO-UHFFFAOYSA-N
Synonym n-benzylidenemethylamine, n-benzylidenemethanamine, benzylidenemethylamine, n-methylbenzaldimine, methanamine, n-phenylmethylene, benzylidenemethanamine, n-methylbenzylideneamine, methylamine, n-benzylidene, n-phenylmethylene methanamine, benzylidene-methyl-amine
PubChem CID 73954
IUPAC Name N-methyl-1-phenylmethanimine
SMILES CN=CC1=CC=CC=C1

Specifications

Density 0.967
Boiling Point 66°C to 69°C (10 mmHg)
Flash Point 77°C (170°F)
Refractive Index 1.552
Quantity 25 g
Beilstein 878278
Formula Weight 119.17
Percent Purity ≥98%
Chemical Name or Material N-Benzylidenemethylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-728-2
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.