Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyliminodiacetic Acid 98.0+%, TCI America™
SDP

Catalog No. B136325G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 3987-53-9
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.228
MDL Number MFCD00004285
InChI Key SZQUPQVVCLFZLC-UHFFFAOYSA-N
PubChem CID 77606
IUPAC Name 2-[benzyl(carboxymethyl)amino]acetic acid
SMILES C1=CC=C(C=C1)CN(CC(=O)O)CC(=O)O

Specifications

Melting Point 201°C
Color White
Quantity 25 g
Formula Weight 223.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzyliminodiacetic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.