Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester 1-(N-succinimidyl) ester, 95%

Click to view available options
Quantity:
250mg
1g
5g
3338-32-7
C20H24N2O8
420.418
MFCD00153328
JUMSBOKRGDETHL-AWEZNQCLSA-N
z-asp otbu-osu,s-4-tert-butyl 1-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino succinate,z-asp obut-osu,n-cbz-,a-t-butyl-l-aspartic acid n-hydroxysuccinimide ester,z-l-aspartic acid 4-tert-butyl-1-n-succinimidyl ester,n-benzyloxycarbonyl-l-aspartic acid 1-succinimidyl 4-tert-butyl ester,s-4-tert-butyl 1-2,5-dioxopyrrolidin-1-yl-2-benzyloxy carbonyl amino succinate,n-alpha-benzyloxycarbonyl-l-aspartic acid alpha-n-hydroxysuccinimidyl beta-t-butyl ester,tert-butyl 2,5-dioxoazolidinyl 2s-2-phenylmethoxy carbonylamino butane-1,4-dioate,1-tert-butyl 2,5-dioxopyrrolidin-1-yl 3s-3-benzyloxy carbonyl amino butanedioate
11026196
4-O-tert-butyl 1-O-(2,5-dioxopyrrolidin-1-yl) (2S)-2-(phenylmethoxycarbonylamino)butanedioate
CC(C)(C)OC(=O)CC(C(=O)ON1C(=O)CCC1=O)NC(=O)OCC2=CC=CC=C2
3338-32-7
C20H24N2O8
420.418
MFCD00153328
JUMSBOKRGDETHL-AWEZNQCLSA-N
z-asp otbu-osu,s-4-tert-butyl 1-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino succinate,z-asp obut-osu,n-cbz-,a-t-butyl-l-aspartic acid n-hydroxysuccinimide ester,z-l-aspartic acid 4-tert-butyl-1-n-succinimidyl ester,n-benzyloxycarbonyl-l-aspartic acid 1-succinimidyl 4-tert-butyl ester,s-4-tert-butyl 1-2,5-dioxopyrrolidin-1-yl-2-benzyloxy carbonyl amino succinate,n-alpha-benzyloxycarbonyl-l-aspartic acid alpha-n-hydroxysuccinimidyl beta-t-butyl ester,tert-butyl 2,5-dioxoazolidinyl 2s-2-phenylmethoxy carbonylamino butane-1,4-dioate,1-tert-butyl 2,5-dioxopyrrolidin-1-yl 3s-3-benzyloxy carbonyl amino butanedioate
11026196
4-O-tert-butyl 1-O-(2,5-dioxopyrrolidin-1-yl) (2S)-2-(phenylmethoxycarbonylamino)butanedioate
CC(C)(C)OC(=O)CC(C(=O)ON1C(=O)CCC1=O)NC(=O)OCC2=CC=CC=C2
Beilstein 1558119
Formula Weight 420.42
Physical Form Powder
Percent Purity 95%
Color White
Melting Point 148°C to 152°C
Quantity 1g
Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester 1-(N-succinimidyl) ester

storageNote1 : 2° to 8°C