Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-N-methyl-L-isoleucine, 95%

Click to view available options
Quantity:
250mg
1g
5g
42417-66-3
C15H21NO4
279.336
MFCD00235876
QUMDFJSXQJTNDA-AAEUAGOBSA-N
z-n-me-ile-oh,n-cbz-n-methyl-l-isoleucine,2s,3s-2-benzyloxy carbonyl methyl amino-3-methylpentanoic acid,cbz-n-me-ile-oh,z-n-me-ile-oh ?,benzyloxycarbonyl-n-alpha-methyl-l-isoleucine,n-benzyloxy carbonyl-n-methyl-l-isoleucine,2s,3s-2-benzyloxy carbonyl methyl amino-3-methylpentanoic acid,n-benzyloxy carbonyl-n-methyl-l-isoleucine,ambotzzaa1014
7019518
(2S,3S)-3-methyl-2-[methyl(phenylmethoxycarbonyl)amino]pentanoic acid
CCC(C)C(C(=O)O)N(C)C(=O)OCC1=CC=CC=C1
42417-66-3
C15H21NO4
279.336
MFCD00235876
QUMDFJSXQJTNDA-AAEUAGOBSA-N
z-n-me-ile-oh,n-cbz-n-methyl-l-isoleucine,2s,3s-2-benzyloxy carbonyl methyl amino-3-methylpentanoic acid,cbz-n-me-ile-oh,z-n-me-ile-oh ?,benzyloxycarbonyl-n-alpha-methyl-l-isoleucine,n-benzyloxy carbonyl-n-methyl-l-isoleucine,2s,3s-2-benzyloxy carbonyl methyl amino-3-methylpentanoic acid,n-benzyloxy carbonyl-n-methyl-l-isoleucine,ambotzzaa1014
7019518
(2S,3S)-3-methyl-2-[methyl(phenylmethoxycarbonyl)amino]pentanoic acid
CCC(C)C(C(=O)O)N(C)C(=O)OCC1=CC=CC=C1
Beilstein 6589752
Formula Weight 279.34
Physical Form Powder
Percent Purity 95%
Color White
Quantity 250mg
Chemical Name or Material N-Benzyloxycarbonyl-N-methyl-L-isoleucine

storageNote1 : 2° to 8°C