Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N″-Biotinyl-6-aminohexanoylhydrazide 97.0+%, TCI America™
SDP

Catalog No. H1071100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1071100MG 100 mg
H107125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1071100MG Supplier TCI America Supplier No. H1071100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 109276-34-8
Molecular Formula C16H29N5O3S
Molecular Weight (g/mol) 371.5
MDL Number MFCD00077659
InChI Key IJJWOSAXNHWBPR-HUBLWGQQSA-N
Synonym Biotin-LC-Hydrazide, N-(6-Hydrazinyl-6-oxohexyl)biotinamide
PubChem CID 83900
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-(6-hydrazinyl-6-oxohexyl)pentanamide
SMILES C1C2C(C(S1)CCCCC(=O)NCCCCCC(=O)NN)NC(=O)N2

Specifications

Melting Point 211°C
Color White
Quantity 100 mg
Formula Weight 371.50
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N′′-Biotinyl-6-aminohexanoylhydrazide
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.