Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-1-(p-toluenesulfonyl)-L-histidine dicyclohexylammonium salt, 98%, Thermo Scientific™

Catalog No. AAH6220506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6220506 5 g
AAH6220514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6220506 Supplier Thermo Scientific Chemicals Supplier No. H6220506

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65057-34-3
Molecular Formula C30H46N4O6S
Molecular Weight (g/mol) 590.78
MDL Number MFCD00039005
InChI Key RNRUVXUHYUOWJW-RSAXXLAASA-N
Synonym boc-his tos-oh.dcha, dicyclohexylamine s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoate, boc-his toc-oh.dcha, n-tert-butoxycarbonyl-1-p-tosyl-l-histidine, compound with dicyclohexylamine 1:1, n-alpha-t-butyloxycarbonyl-n-im-t-tosyl-l-histidine dicyclohexylamine, boc-his toc-oh?cha, 2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid; dicha, l-histidine, n-1,1-dimethylethoxy carbonyl-1-4-methylphenyl sulfonyl-, compd with n-cyclohexylcyclohexanamine 1:1, n-cyclohexylcyclohexanamine; 2s-3-1-4-methylphenyl sulfonylimidazol-4-yl-2-tert-butoxycarbonylamino propanoic acid
PubChem CID 6455009
SMILES C1CCC(CC1)NC1CCCCC1.CC1=CC=C(C=C1)S(=O)(=O)N1C=NC(C[C@H](NC(=O)OC(C)(C)C)C(O)=O)=C1

Specifications

IUPAC Name N-cyclohexylcyclohexanamine;(2S)-3-[1-(4-methylphenyl)sulfonylimidazol-4-yl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Quantity 5 g
Formula Weight 590.77
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-1-(p-toluenesulfonyl)-L-histidine dicyclohexylammonium salt
EINECSNumber 265-328-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.