Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-3-dimethylamino-L-alanine, 97%

Catalog No. AAH5281003
Click to view available options
Quantity:
250 mg
1 g

CAS: 94778-71-9 | C10H20N2O4 | 232.28 g/mol

N-Boc-3-dimethylamino-L-alanine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-3-dimethylamino-L-alanine is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94778-71-9
Molecular Formula C10H20N2O4
Molecular Weight (g/mol) 232.28
MDL Number MFCD02682423
InChI Key VCDQZVYJKDSORW-ZETCQYMHSA-N
Synonym boc-aza-l-leucine, n-tert-butoxycarbonyl-3-dimethylamino-l-alanine, 2s-2-tert-butoxycarbonyl amino-3-dimethylamino propanoic acid, boc-, ?-n,n-dimethylamino-l-ala, boc-beta-n,n-dimethylamino-l-ala, na-boc-s-2-amino-3-dimethylamino propionic acid, n-alpha-boc-s-2-amino-3-dimethylamino propionic acid, s-2-tert-butoxycarbonylamino-3-dimethylamino-propionic acid, l-alanine,3-dimethylamino-n-1,1-dimethylethoxy carbonyl, 2s-2-tert-butoxy carbonyl amino-3-dimethylamino propanoic acid
PubChem CID 17998956
IUPAC Name (2S)-3-(dimethylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CN(C)C)C(=O)O

Specifications

Quantity 1 g
Formula Weight 232.28
Percent Purity 97%
Chemical Name or Material N-Boc-3-dimethylamino-L-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.