Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-3-hydroxy-L-valine, 97%

Catalog No. AAH5242903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5242903 1 g
AAH52429MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5242903 Supplier Thermo Scientific Chemicals Supplier No. H5242903
Add to Cart

CAS: 102507-13-1 | C10H19NO5 | 233.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102507-13-1
Molecular Formula C10H19NO5
Molecular Weight (g/mol) 233.26
MDL Number MFCD03094792
InChI Key SZVRVSZFEDIMFM-ZCFIWIBFSA-N
Synonym s-2-tert-butoxycarbonyl amino-3-hydroxy-3-methylbutanoic acid, n-boc-s-2-amino-3-hydroxy-3-methylbutanoic acid, s-2-n-boc-amino-3-hydroxy-3-methylbutyric acid, s-n-boc-2-amino-3-hydroxy-3-methylbutanoic acid, n-tert-butoxycarbonyl-3-hydroxy-l-valine, 2s-2-n-tert-butoxycarbonyl amino-3-hydroxy-3-methylbutanoic acid, n-boc-2-amino-3-hydroxy-3-methylbutanoic acid, s-2-boc-amino-3-hydroxy-3-methylbutyric acid, boc-s-2-amino-3-hydroxy-3-methylbutanoic acid, l-valine, n-1,1-dimethylethoxy carbonyl-3-hydroxy
PubChem CID 11075314
IUPAC Name (2S)-3-hydroxy-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)O

Specifications

Quantity 1 g
Formula Weight 233.26
Percent Purity 97%
Chemical Name or Material N-Boc-3-hydroxy-L-valine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.