Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-4-oxo-L-proline methyl ester, 97%, Thermo Scientific Chemicals

Catalog No. AAH2778206
Click to view available options
Quantity:
250 mg
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102195-80-2
Molecular Formula C11H17NO5
Molecular Weight (g/mol) 243.259
MDL Number MFCD01861778
InChI Key UPBHYYJZVWZCOZ-QMMMGPOBSA-N
Synonym boc-4-oxo-pro-ome, n-boc-4-oxo-l-proline methyl ester, s-1-tert-butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, 2s-1-boc-4-oxo-proline methyl ester, methyl 1-boc-4-oxo-proline, 1-boc-4-oxo-l-proline methyl ester, tert-butyl 2s-2-methoxycarbonyl-4-oxopyrrolidine-1-carboxylate, 2s-4-oxo-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid methyl ester, 1,2-pyrrolidinedicarboxylic acid, 4-oxo-, 1-1,1-dimethylethyl 2-methyl ester, 2s
PubChem CID 10988485
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S)-4-oxopyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CC(=O)CC1C(=O)OC

Specifications

Melting Point 35°C to 40°C
Quantity 5 g
Formula Weight 243.26
Percent Purity 97%
Chemical Name or Material N-Boc-4-oxo-L-proline methyl ester
Hazard Category H302+H312+H332-H318
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.