Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-aniline, 97%

Catalog No. p-7052670
Click to view available options
Quantity:
5 g
25 g

CAS: 1-3-3422 | C11H15NO2 | 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1-3-3422
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.25
MDL Number MFCD00043723
InChI Key KZZHPWMVEVZEFG-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NC1=CC=CC=C1

Specifications

Melting Point 133°C to 137°C
Color Brown to White
Linear Formula C6H5NHCO2C(CH3)3
Quantity 5 g
Synonym n-boc-aniline, tert-butyl phenylcarbamate, n-boc aniline, t-butyl carbanilate, tert-butyl carbanilate, carbanilic acid, tert-butyl ester, carbamic acid, phenyl-, 1,1-dimethylethyl ester, n-tert-butoxycarbonyl aniline, tert-butyl n-phenylurethane, carbanilic acid tert-butyl ester
IUPAC Name tert-butyl N-phenylcarbamate
PubChem CID 137930
Formula Weight 193.25
Percent Purity 0.97
Physical Form Solid
Chemical Name or Material N-Boc-aniline
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.