Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-beta-phenyl-D-phenylalanine, 98%

Catalog No. AAH5688503
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5688503 1 g
AAH56885ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5688503 Supplier Thermo Scientific Chemicals Supplier No. H5688503
Add to Cart

CAS: 143060-31-5 | C20H23NO4 | 341.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143060-31-5
Molecular Formula C20H23NO4
Molecular Weight (g/mol) 341.41
MDL Number MFCD00191187
InChI Key TYJDOLCFYZSNQC-UHFFFAOYNA-N
Synonym boc-d-3,3-diphenylalanine, boc-d-ala 3,3-diphenyl-oh, r-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, boc-3,3-diphenyl-d-alanine, r-n-boc-2-amino-3,3-diphenylpropionic acid, n-boc-beta-phenyl-d-phenylalanine, n-tert-butoxycarbonyl-beta-phenyl-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, n-tert-butoxycarbonyl-3-phenyl-l-phenylalanine, n-boc-b-phenyl-d-phenylalanine
PubChem CID 7019136
SMILES CC(C)(C)OC(=O)NC(C(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 157°C
Quantity 1 g
Optical Rotation −30 (c=1 in Ethanol)
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3,3-diphenylpropanoic acid
Formula Weight 341.4
Percent Purity 98%
Chemical Name or Material N-Boc-beta-phenyl-D-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.