Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nα-BOC-D-Arginine hydrochloride hydrate, 99%, Thermo Scientific™

Catalog No. AC275730025
Click to view available options
Quantity:
2.5 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114622-81-0
Molecular Formula C11H25ClN4O5
Molecular Weight (g/mol) 328.794
MDL Number MFCD00065554
InChI Key OVXLPYFDJUFEHQ-KLXURFKVSA-N
Synonym boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, boc-arg-oh.hcl.h2o, boc-d-arg-oh hcl h2o, boc-arg-oh hcl h2o, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, a-boc-d-arginine hydrochloride monohydrate, pubchem14932, boc-l-arg-ohhclh2o
PubChem CID 2729004
IUPAC Name (2S)-5-(diaminomethylideneamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate;hydrochloride
SMILES CC(C)(C)OC(=O)NC(CCCN=C(N)N)C(=O)O.O.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula HN=C(NH2)nH(CH2)3CH(COOH)nHCO2C(CH3)3
Specific Rotation +7.5° to +9.5° (20°C, 589nm) (c=2, H2O)
Quantity 2.5 g
Formula Weight 310.77
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material Nα-BOC-D-Arginine hydrochloride hydrate, 99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.