Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-D-aspartic acid 4-benzyl ester, 98%

Click to view available options
Quantity:
1g
5g
25g
51186-58-4
C16H21NO6
323.345
MFCD00038255
SOHLZANWVLCPHK-GFCCVEGCSA-N
boc-d-asp obzl-oh,boc-d-aspartic acid 4-benzyl ester,r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid,2r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid,n-alpha-t-boc-d-aspartic acid beta-benzyl ester,boc-d-asparticacid4-benzylester,pubchem11179,boc-d-asp obn-oh,boc-d-aspartic acid b-benzyl ester,boc-d-aspartic acid ?-benzyl ester
6994663
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O
51186-58-4
C16H21NO6
323.345
MFCD00038255
SOHLZANWVLCPHK-GFCCVEGCSA-N
boc-d-asp obzl-oh,boc-d-aspartic acid 4-benzyl ester,r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid,2r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid,n-alpha-t-boc-d-aspartic acid beta-benzyl ester,boc-d-asparticacid4-benzylester,pubchem11179,boc-d-asp obn-oh,boc-d-aspartic acid b-benzyl ester,boc-d-aspartic acid ?-benzyl ester
6994663
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O
Applications
N-Boc-D-aspartic acid 4-benzyl ester is used as an intermediate in organic synthesis and pharmaceuticals.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Quantity 5g
Beilstein 2305471
Formula Weight 323.34
Percent Purity 98%
Chemical Name or Material N-Boc-D-aspartic acid 4-benzyl ester

TSCA : No

Recommended Storage : Ambient temperatures