Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-D-aspartic acid, 98%, Thermo Scientific™

Catalog No. AAH6282003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6282003 1 g
AAH6282006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6282003 Supplier Thermo Scientific Chemicals Supplier No. H6282003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62396-48-9
Molecular Formula C9H15NO6
Molecular Weight (g/mol) 233.22
MDL Number MFCD00798618
InChI Key KAJBMCZQVSQJDE-RXMQYKEDSA-N
Synonym boc-d-aspartic acid, boc-d-asp-oh, n-boc-d-aspartic acid, n-tert-butoxycarbonyl-d-aspartic acid, d-aspartic acid, n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino butanedioic acid, boc-d-asparticacid, pubchem12159, n-t-boc-d-aspartic acid, ksc494c3p
PubChem CID 7021110
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)O

Specifications

Quantity 1 g
Formula Weight 233.22
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-D-aspartic acid
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.