Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-D-Phenylalaninal, 97%, Thermo Scientific™

Catalog No. AC376732500
Click to view available options
Quantity:
250 mg
1 g

Chemical Identifiers

CAS 77119-85-8
Molecular Formula C14H19NO3
Molecular Weight (g/mol) 249.31
MDL Number MFCD00274187
InChI Key ZJTYRNPLVNMVPQ-GFCCVEGCSA-N
Synonym n-boc-d-phenylalaninal, boc-d-phenylalaninal, r-+-2-tert-butoxycarbonylamino-3-phenylpropanal, r-tert-butyl 1-oxo-3-phenylpropan-2-yl carbamate, n-boc-2 r-3-phenylpropanal, tert-butyl n-2r-1-oxo-3-phenylpropan-2-yl carbamate, tert-butyl 2r-1-oxo-3-phenylpropan-2-yl carbamate, carbamic acid, 1r-1-formyl-2-phenylethyl-, 1,1-dimethylethyl ester, boc-d-phe-h, pubchem11217
PubChem CID 7020444
IUPAC Name tert-butyl N-[(2R)-1-oxo-3-phenylpropan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1)C=O

Specifications

Melting Point 85.0°C to 89.0°C
CAS Min % 96.0
CAS Max % 100.0
Color Tan or White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Specific Rotation + 45.00
Quantity 250 mg
Formula Weight 249.31
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-D-Phenylalaninal
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.