Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-D-serine benzyl ester, 95%, Thermo Scientific Chemicals

Catalog No. AAH66514MD
Click to view available options
Quantity:
250 mg
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141527-78-8
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.335
MDL Number MFCD00080265
InChI Key BQADRZHPZVQGCW-GFCCVEGCSA-N
Synonym boc-d-ser-obzl, boc-d-ser-o-bzl, boc-d-serine benzyl ester, r-benzyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, benzyl n-tert-butoxycarbonyl-d-serinate, benzyl 2r-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-d-serine benzylester, boc-d-ser-o-bzl hplc
PubChem CID 12953536
IUPAC Name benzyl (2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CO)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 66°C to 70°C
Color White
Quantity 250 mg
Formula Weight 295.34
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-D-serine benzyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.