Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-D-tyrosine methyl ester, 96%

Catalog No. AAH6682722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6682722 100 g
AAH6682706 5 g
AAH6682714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6682722 Supplier Thermo Scientific Chemicals Supplier No. H6682722

CAS: 76757-90-9 | C15H21NO5 | 295.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76757-90-9
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.34
MDL Number MFCD00057824
InChI Key NQIFXJSLCUJHBB-UHFFFAOYNA-N
Synonym boc-d-tyr-ome, r-methyl 2-tert-butoxycarbonyl amino-3-4-hydroxyphenyl propanoate, boc-d-tyrosine methyl ester, d-tyrosine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-tert-butoxycarbonyl amino-3-4-hydroxyphenyl propanoate, methyl 2r-2-tert-butoxy carbonyl amino-3-4-hydroxyphenyl propanoate, methyl boc-d-tyrosinate, boc-d-tyrosine-methyl ester, n-t-butoxycarbonyl-d-tyrosine methyl ester
PubChem CID 7016055
IUPAC Name methyl 2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
SMILES COC(=O)C(CC1=CC=C(O)C=C1)NC(=O)OC(C)(C)C

Specifications

Color White
Beilstein 4704567
Quantity 100 g
Formula Weight 295.34
Percent Purity 96%
Physical Form Powder
Chemical Name or Material N-Boc-D-tyrosine methyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.