Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-DL-1-Aminoindane-1-carboxylic acid, 99%, Thermo Scientific™

Catalog No. AC302702500
Click to view available options
Quantity:
250 mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214139-26-1
Molecular Formula C15H19NO4
Molecular Weight (g/mol) 277.32
InChI Key UZCCJRDJRMVGLU-UHFFFAOYSA-N
Synonym 1-boc-amino-1-indanecarboxylic acid, 1-tert-butoxycarbonyl amino-2,3-dihydro-1h-indene-1-carboxylic acid, n-boc-d,l-1-aminoindane-1-carboxylic acid, r,s-boc-1-aminoindane-1-carboxylic acid, 1-boc-amino-1-indanecarboxylicacid, 1-tert-butoxycarbonyl amino-2,3-dihydroindene-1-carboxylic acid, boc-r,s-1-aminoindane-1-carboxylic acid, 1-tert-butoxy carbonylamino indanecarboxylic acid, 1-2-methylpropan-2-yl oxycarbonylamino-2,3-dihydroindene-1-carboxylic acid
PubChem CID 2733195
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonylamino]-2,3-dihydroindene-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CCC2=CC=CC=C21)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 156°C to 159°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Quantity 250 mg
Formula Weight 277.32
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-DL-1-Aminoindane-1-carboxylic acid, 99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.