Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC Ethyl oxamate, 98%, Thermo Scientific™

Catalog No. AC333990010
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216959-34-1
Molecular Formula C9H15NO5
Molecular Weight (g/mol) 217.221
MDL Number MFCD01631201
InChI Key DSDQWJVXMDHQLK-UHFFFAOYSA-N
Synonym ethyl n-tert-butoxycarbonyl oxamate, ethyl n-boc-oxamidate, ethyl 2-tert-butoxycarbonyl amino-2-oxoacetate, ethyl n-boc-oxamate, ethyl 2-tert-butoxycarbonylamino-2-oxoacetate, ethyl 2-tert-butoxycarbonylamino-2-oxo-acetate, n-boc ethyl oxamate, ethyl 2-tert-butoxycarbonyl-2-oxoacetate
PubChem CID 2733665
IUPAC Name ethyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]-2-oxoacetate
SMILES CCOC(=O)C(=O)NC(=O)OC(C)(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Refractive Index 1.4490 to 1.4510 (20°C, 589nm)
Quantity 1 g
Formula Weight 217.22
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material N-BOC Ethyl oxamate
Show More Show Less
Hazard Category MOISTURE SENSITIVE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.