Learn More
N-Boc-ethylenediamine, 98%, may cont up to 5% tert-butanol
Description
N-Boc-ethylenediamine is used in the synthesis of thyronamine derivatives. It is also used in the preparation of (2-isothiocyanato-ethyl)-carbamic acid tert-butyl ester by reacting with carbon disulfide. Further, it is used in both oligonucleotide and peptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Boc-ethylenediamine is used in the synthesis of thyronamine derivatives. It is also used in the preparation of (2-isothiocyanato-ethyl)-carbamic acid tert-butyl ester by reacting with carbon disulfide. Further, it is used in both oligonucleotide and peptide synthesis.
Solubility
Miscible with methanol and chloroform. Slightly miscible with water.
Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents and acids.
Specifications
Specifications
| Boiling Point | 72°C (0.1mmHg) |
| Refractive Index | 1.457 |
| UN Number | UN2735 |
| Beilstein | 1932330 |
| Solubility Information | Miscible with methanol and chloroform. Slightly miscible with water. |
| Density | 1.016 |
| Quantity | 5 g |
| Flash Point | >110°C (230°F) |
| Formula Weight | 160.22 |
| Sensitivity | Air Sensitive |
| Show More |
Safety and Handling
| Hazard Category | H314-H335-H336 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S. |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.