Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-L-alaninamide, 96%

Catalog No. AAH6260406
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6260406 5 g
AAH62604MD 250 mg
AAH6260403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6260406 Supplier Thermo Scientific Chemicals Supplier No. H6260406
Add to Cart

CAS: 85642-13-3 | C8H16N2O3 | 188.23 g/mol

N-Boc-L-alaninamide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-alaninamide is used as pharmaceutical intermediate.

Solubility
Soluble in water or 1% acetic acid.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85642-13-3
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.23
MDL Number MFCD03701447
InChI Key GZKKSKKIABUUCX-UHFFFAOYNA-N
Synonym boc-ala-nh2, n-t-butyloxycarbonyl-l-alanine amide, s-tert-butyl 1-amino-1-oxopropan-2-yl carbamate, tert-butyl n-1s-1-carbamoylethyl carbamate, boc-beta-ala-nh2, boc-l-ala-nh2, boc-l-alanine-nh2, boc-l-alanine amide, s-boc-alaninamide, ambotzbaa5310
PubChem CID 7015295
SMILES CC(NC(=O)OC(C)(C)C)C(N)=O

Specifications

Quantity 5 g
Solubility Information Soluble in water or 1% acetic acid.
IUPAC Name tert-butyl N-(1-carbamoylethyl)carbamate
Formula Weight 188.22
Percent Purity 96%
Physical Form Solid
Chemical Name or Material N-Boc-L-alaninamide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.