Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Boc-L-phenylalanine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
N-Boc-L-phenylalanine is a derivative of Phenylalanine used in enantioselective hydrolysis of amino acid esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-L-phenylalanine is a derivative of Phenylalanine used in enantioselective hydrolysis of amino acid esters.
Solubility
Soluble in methanol, dichloromethane, dimethylformamide and N-methyl-2-pyrrolidone.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
N-Boc-L-phenylalanine is a derivative of Phenylalanine used in enantioselective hydrolysis of amino acid esters.
Solubility
Soluble in methanol, dichloromethane, dimethylformamide and N-methyl-2-pyrrolidone.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Melting Point | 84°C to 88°C |
Quantity | 5 g |
Beilstein | 2219729 |
Solubility Information | Soluble in methanol,dichloromethane,dimethylformamide and N-methyl-2-pyrrolidone. |
Optical Rotation | +25° (c=1 in Ethanol) |
IUPAC Name | 2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoic acid |
Formula Weight | 265.31 |
Percent Purity | 99% |
Chemical Name or Material | N-Boc-L-phenylalanine |
Safety and Handling
EINECSNumber | 237-305-5 |
TSCA | Yes |
Recommended Storage | Keep cold |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction