Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-L-phenylalaninol, 99%

Catalog No. p-7027943
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 66605-57-0 | C14H21NO3 | 251.33 g/mol

N-Boc-L-phenylalaninol is employed in the synthesis of biochemically active compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-phenylalaninol is employed in the synthesis of biochemically active compounds.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66605-57-0
Molecular Formula C14H21NO3
Molecular Weight (g/mol) 251.33
MDL Number MFCD00076976
InChI Key LDKDMDVMMCXTMO-LBPRGKRZSA-N
Synonym n-boc-l-phenylalaninol, boc-l-phenylalaninol, boc-phenylalaninol, s-2-boc-amino-3-phenyl-1-propanol, n-tert-butoxycarbonyl-l-phenylalaninol, tert-butyl n-2s-1-hydroxy-3-phenylpropan-2-yl carbamate, s-tert-butyl 1-hydroxy-3-phenylpropan-2-yl carbamate, s-n-tert-butoxycarbonyl-phenylalaninol, s---2-tert-butoxycarbonylamino-3-phenyl-1-propanol
PubChem CID 2733675
IUPAC Name tert-butyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)N[C@H](CO)CC1=CC=CC=C1

Specifications

Melting Point 95°C to 98°C
Beilstein 3652004
Solubility Information Insoluble in water.
Quantity 1 g
Optical Rotation −27.5° ± 2.5° (c=1 in Methanol)
Formula Weight 251.33
Percent Purity 99%
Chemical Name or Material N-Boc-L-phenylalaninol
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.