Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-L-tryptophanol, 95%

Click to view available options
Quantity:
250mg
1g
5g
82689-19-8
C16H22N2O3
290.363
MFCD00235953
JEFQUFUAEKORKL-LBPRGKRZSA-N
n-boc-l-tryptophanol,boc-tryptophanol,boc-l-tryptophanol,boc-trp-ol,s-tert-butyl 1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate,s-3-2-boc-amino-3-hydroxypropyl-indole,boc-d-trp-ol,n-alpha-boc-l-tryptophanol,boc-trypotophanol,boc-l-trp-ol
7019533
tert-butyl N-[(2S)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate
CC(C)(C)OC(=O)NC(CC1=CNC2=CC=CC=C21)CO
82689-19-8
C16H22N2O3
290.363
MFCD00235953
JEFQUFUAEKORKL-LBPRGKRZSA-N
n-boc-l-tryptophanol,boc-tryptophanol,boc-l-tryptophanol,boc-trp-ol,s-tert-butyl 1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate,s-3-2-boc-amino-3-hydroxypropyl-indole,boc-d-trp-ol,n-alpha-boc-l-tryptophanol,boc-trypotophanol,boc-l-trp-ol
7019533
tert-butyl N-[(2S)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate
CC(C)(C)OC(=O)NC(CC1=CNC2=CC=CC=C21)CO
Formula Weight 290.36
Physical Form Solid
Percent Purity 95%
Melting Point 118°C to 122°C
Quantity 250mg
Chemical Name or Material N-Boc-L-tryptophanol