Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-L-valine N-succinimidyl ester, 97%

Catalog No. aah6655922
Click to view available options
Quantity:
25 g
100 g

CAS: 3392-12-9 | C14H22N2O6 | 314.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3392-12-9
Molecular Formula C14H22N2O6
Molecular Weight (g/mol) 314.34
MDL Number MFCD00037906
InChI Key POBDBYGSGKMZPH-NSHDSACASA-N
IUPAC Name 2,5-dioxopyrrolidin-1-yl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-methylbutanoate
SMILES CC(C)[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O

Specifications

Color White
Beilstein 1551859
Synonym boc-val-osu, boc-l-valine hydroxysuccinimide ester, boc-l-val-osu, s-2,5-dioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino-3-methylbutanoate, tert-butyl s-1-2,5-dioxopyrrolidin-1-yl oxy carbonyl-2-methylpropyl carbamate, 2,5-dioxopyrrolidin-1-yl n-tert-butoxycarbonyl-l-valinate, bocvalosu, 2,5-dioxopyrrolidin-1-yl 2s-2-tert-butoxycarbonyl amino-3-methylbutanoate, t-boc-val-osu, pubchem20978
Quantity 100 g
PubChem CID 98768
Formula Weight 314.34
Sensitivity Moisture sensitive
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-Boc-L-valine N-succinimidyl ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-236-5
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.