Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-N-methyl-1,3-diaminopropane, 95%, Thermo Scientific Chemicals

Catalog No. AAH66932MD
Click to view available options
Quantity:
250 mg
1 g

It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150349-36-3
Molecular Formula C9H20N2O2
Molecular Weight (g/mol) 188.271
MDL Number MFCD01631189
InChI Key PNQYAMWGTGWJDW-UHFFFAOYSA-N
Synonym n-3-aminopropyl-n-methylcarbamic acid tert-butyl ester, tert-butyl n-3-aminopropyl-n-methylcarbamate, tert-butyl 3-aminopropyl methyl carbamate, 3-n-boc-n-methylamino propylamine, n-boc-n-methyl-1,3-diaminopropane, n-3-amino-propyl-n-methylcarbamic acid t-butyl ester, acid tert-butyl ester, 3-n-tert-butoxycarbonyl-n-methylamino propylamine, tert-butyl n-3-aminopropyl-n-methyl carbamate
PubChem CID 2756543
IUPAC Name tert-butyl N-(3-aminopropyl)-N-methylcarbamate
SMILES CC(C)(C)OC(=O)N(C)CCCN

Specifications

Color Yellow
Solubility Information Slightly soluble in water.
Quantity 250 mg
Formula Weight 188.27
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-Boc-N-methyl-1,3-diaminopropane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.