Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-N-methyl-L-serine, 97%

Catalog No. aah63314md
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 101772-29-6 | C9H17NO5 | 219.237 g/mol

N-Boc-N-methyl-L-serine is used as a precursor in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-N-methyl-L-serine is used as a precursor in organic synthesis and pharmaceuticals.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101772-29-6
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.237
MDL Number MFCD00037249
InChI Key NJQGIDVCNBZXLG-LURJTMIESA-N
Synonym boc-n-me-ser-oh, boc-n-methyl-l-serine, boc-n-a-methyl-l-serine, boc-n-, a-methyl-l-serine, s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, n-boc-n-methyl-l-serine, l-serine,n-1,1-dimethylethoxy carbonyl-n-methyl, boc-meser-oh, 2s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, boc-l-meser-oh
PubChem CID 7009594
IUPAC Name (2S)-3-hydroxy-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
SMILES CC(C)(C)OC(=O)N(C)C(CO)C(=O)O

Specifications

Melting Point 83°C to 87°C
Beilstein 8685334
Quantity 250 mg
Formula Weight 219.23
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-N-methyl-L-serine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.