Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-O-benzyl-L-tyrosine, 98%, Thermo Scientific Chemicals

Catalog No. AAB2345506
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2130-96-3
Molecular Formula C21H25NO5
Molecular Weight (g/mol) 371.43
MDL Number MFCD00065597
InChI Key ZAVSPTOJKOFMTA-LGWFVXIRNA-N
Synonym boc-tyr bzl-oh, boc-o-benzyl-l-tyrosine, n-boc-o-benzyl-l-tyrosine, n-tert-butoxycarbonyl-o-benzyl-l-tyrosine, s-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-butoxycarbonyl-o-benzyl-l-tyrisine, o-benzyl-n-tert-butoxycarbonyl-l-tyrosine, 2s-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, o-benzyl-n-tert-butoxy carbonyl-l-tyrosine, tyrosine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl
PubChem CID 98763
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylmethoxyphenyl)propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O

Specifications

Melting Point 108°C to 110°C
Quantity 5 g
Formula Weight 371.43
Percent Purity 98%
Chemical Name or Material N-Boc-O-benzyl-L-tyrosine
EINECSNumber 218-349-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.