Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-O-methyl-L-serine, 97%

Catalog No. p-7035120
Click to view available options
Quantity:
250 mg
1 g

CAS: 51293-47-1 | C9H17NO5 | 219.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51293-47-1
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.237
MDL Number MFCD00153314
InChI Key RFGMSGRWQUMJIR-LURJTMIESA-N
Synonym boc-o-methyl-l-serine, boc-ser me-oh, s-2-tert-butoxycarbonyl amino-3-methoxypropanoic acid, boc-ser me-oh.dcha, n-boc-o-methyl-l-serine, s-2-tert-butoxycarbonylamino-3-methoxypropanoic acid, boc-ser me-oh dcha, boc-o-methyl-l-ser, serine, n-1,1-dimethylethoxy carbonyl-o-methyl, 2s-2-tert-butoxycarbonyl amino-3-methoxypropanoic acid
PubChem CID 7016354
IUPAC Name (2S)-3-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(COC)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 219.24
Percent Purity 97%
Chemical Name or Material N-Boc-O-methyl-L-serine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.