Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-S-trityl-L-cysteine, 97%

Catalog No. p-7039960
Click to view available options
Quantity:
5 g
25 g

CAS: 21947-98-8 | C27H29NO4S | 463.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21947-98-8
Molecular Formula C27H29NO4S
Molecular Weight (g/mol) 463.592
MDL Number MFCD00038251
InChI Key JDTOWOURWBDELG-QHCPKHFHSA-N
Synonym boc-cys trt-oh, n-boc-s-trityl-l-cysteine, boc-s-trityl-l-cysteine, n-tert-butoxycarbonyl-s-trityl-l-cysteine, r-2-tert-butoxycarbonyl amino-3-tritylthio propanoic acid, boc-cysteine trt-oh, 2r-2-tert-butoxy carbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-triphenylmethyl, boc-l-cysteine trityl, 2r-2-tert-butoxycarbonyl amino-3-triphenylmethyl sulfanyl propanoic acid
PubChem CID 11167161
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-tritylsulfanylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O

Specifications

Melting Point 143°C to 146°C
Quantity 5 g
Beilstein 2028965
Optical Rotation +28° (c=1 in Ethanol)
Formula Weight 463.59
Percent Purity 97%
Chemical Name or Material N-Boc-S-trityl-L-cysteine
EINECSNumber 244-674-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.