Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-trans-4-(Fmoc-amino)-L-proline, 97%, Thermo Scientific™

Catalog No. AAH6687803
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 176486-63-8
Molecular Formula C25H28N2O6
Molecular Weight (g/mol) 452.507
MDL Number MFCD00673783
InChI Key UPXRTVAIJMUAQR-VFNWGFHPSA-N
Synonym 2s,4r-4-9h-fluoren-9-yl methoxy carbonyl amino-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-boc-trans-4-n-fmoc-amino-l-proline, 2s,4r-n-tert-butoxycarbonyl-4-n-9-fluorenylmethoxycarbonyl aminopyrrolidine-2-carboxylic acid, 2s,4r-4-9h-fluoren-9-yl methoxy carbonylamino-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, 2s,4r-1-tert-butoxycarbonyl-4-9h-fluoren-9-ylmethoxy carbonyl amino pyrrolidine-2-carboxylic acid, ambotzbaa1783, 2s,4r-fmoc-4-amino-1-boc-pyrrolidine-2-carboxyli, 2s,4r-1-boc-4-n-fmoc-4-aminopyrrolidine-2-carboxylic acid, 2s,4r-trans-n-alpha-t-butyloxycarbonyl-4-9-fluorenylmethyloxycarbonyl amino-l-proline
PubChem CID 2756135
IUPAC Name (2S,4R)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point 197°C to 201°C
Color White
Quantity 1 g
Formula Weight 452.5
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-Boc-trans-4-(Fmoc-amino)-L-proline
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.