Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Bromophthalimide 95.0+%, TCI America™
SDP

Catalog No. B169725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B169725G 25 g
B16975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B169725G Supplier TCI America Supplier No. B169725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2439-85-2
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.029
MDL Number MFCD00005888
InChI Key MARXMDRWROUXMD-UHFFFAOYSA-N
PubChem CID 75542
IUPAC Name 2-bromoisoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)Br

Specifications

Melting Point 207°C
Quantity 25 g
Synonym n-bromophthalimide, 2-bromoisoindoline-1,3-dione, 2-bromo-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 2-bromo, n-bromophtalimide, n-bromo phthalimide, acmc-1cjlt, 2-bromobenzo c azoline-1,3-dione, 1h-isoindole-1,3 2h-dione,2-bromo
Formula Weight 226.03
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Bromophthalimide
EINECSNumber (5)-0080
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.