Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tert-butoxide, 98+%, pure

Catalog No. AC168881000
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle

CAS: 865-47-4 | C4H9KO | 112.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-47-4
Molecular Formula C4H9KO
Molecular Weight (g/mol) 112.21
InChI Key LPNYRYFBWFDTMA-UHFFFAOYSA-N
Synonym potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
PubChem CID 23665647
IUPAC Name potassium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].[K+]

Specifications

Melting Point 240.0°C to 258.0°C
Color White
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula (CH3)3COK
Quantity 100 g
Fieser 01,911; 02,336; 03,233; 04,399; 05,544; 06,477; 08,407; 09,380; 10,323; 11,432; 12,97; 14,264; 17,289
Solubility Information Solubility in water: reacts with water. Other solubilities: soluble in methanol, ethanol, slightly soluble in diethyl ether, benzene
Formula Weight 112.21
Percent Purity 98+%
Grade Pure
Physical Form Powder
Chemical Name or Material Potassium tert-butoxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
Self-heating in large quantities; may catch fire.
Reacts violently with water.
missing translation for 'healthHazard3' GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several min
EINECSNumber 212-740-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.