Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyl Acetate, ACS, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18611905 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly-Coated Glass Bottle
Steel Can

Catalog No. 18-611-905

Supplier: Spectrum Chemical Manufacturing Corporation B11821LTGL

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 L
Residue after Evaporation 1e-05
Percent Purity 0.995
Grade ACS
Titratable Acid 0.0016 meq/g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.