Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamic acid dimethyl ester hydrochloride, 99%, pure

Catalog No. AC441940500
Click to view available options
Quantity:
10 g
50 g

CAS: 23150-65-4 | C7H13ClNO4 | 210.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23150-65-4
Molecular Formula C7H13ClNO4
Molecular Weight (g/mol) 210.63
MDL Number MFCD00038879
InChI Key BPHCSYSXOKTCMA-UHFFFAOYNA-N
Synonym l-glutamic acid dimethyl ester hydrochloride, h-glu ome-ome.hcl, dimethyl l-glutamate hydrochloride, s-dimethyl 2-aminopentanedioate hydrochloride, unii-185vc5x3pf, 1,5-dimethyl 2s-2-aminopentanedioate hydrochloride, h-glu ome-ome inverted exclamation mark currencyhcl, s-dimethyl 2-aminopentanedioate hcl, glutamic acid dimethyl ester hydrochloride, dimethyl glutamic acid hydrochloride
PubChem CID 12917568
IUPAC Name dimethyl (2S)-2-aminopentanedioate;hydrochloride
SMILES [Cl].COC(=O)CCC(N)C(=O)OC

Specifications

Melting Point 91°C to 93°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Quantity 50 g
Specific Rotation 26
Formula Weight 211.65
Percent Purity 99%
Grade Pure
Physical Form Powder
Chemical Name or Material L-Glutamic acid dimethyl ester hydrochloride
Show More Show Less
EINECSNumber 245-461-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.