Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyl acetate, HPLC Grade, 99.5+%

Catalog No. p-7050647
Click to view available options
Quantity:
250 mL
1 L
4 L
4 x 1 L

CAS: 123-86-4 | C6H12O2 | 116.16 g/mol

n-Butyl acetate is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Melting Point -78°C
Color Colorless
Density 0.881 g/mL
pH 5
Boiling Point 124°C to 126°C
Flash Point 25°C (77°F)
Odor Fruit-like
Refractive Index 1.3945
Linear Formula CH3CO2(CH2)3CH3
Quantity 250 mL
UN Number UN1123
Beilstein 1741921
Merck Index 14,1535
Solubility Information Miscible with ethanol,ethyl ether,alcohol,acetone,chloroform and most hydrocarbonsSlightly miscible with water.
Formula Weight 116.16
Percent Purity ≥99.5%
Grade HPLC
Physical Form Liquid
Chemical Name or Material n-Butyl acetate
Show More Show Less
Hazard Category H226-H336
Hazard Statement GHS H Statement
H226-H336
Flammable liquid and vapor.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYL ACETATES
EINECSNumber 204-658-1
RTECSNumber AF350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.