Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyl butyrate, 99%

Catalog No. p-7027551
Click to view available options
Quantity:
1000 mL

CAS: 109-21-7 | C8H16O2 | 144.21 g/mol

It is very important for the food and beverages industries and used as perfuming agents. It is a applied as a solvent for resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is very important for the food and beverages industries and used as perfuming agents. It is a applied as a solvent for resins.

Solubility
Soluble in water. (1 g/L).

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-21-7
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00009450
InChI Key XUPYJHCZDLZNFP-UHFFFAOYSA-N
Synonym butyl butyrate, n-butyl butyrate, butanoic acid, butyl ester, n-butyl butanoate, 1-butyl butyrate, butyl butylate, butyric acid butyl ester, n-butyl n-butyrate, butyric acid, butyl ester, butyl n-butyrate
PubChem CID 7983
ChEBI CHEBI:87429
IUPAC Name butyl butanoate
SMILES CCCCOC(=O)CCC

Specifications

Melting Point -92°C
Density 0.871
Boiling Point 164°C to 165°C
Flash Point 49°C (120°F)
Refractive Index 1.406
Linear Formula CH3CH2CH2CO2(CH2)3CH3
Quantity 1000 mL
UN Number UN3272
Beilstein 1747101
Merck Index 14,1556
Solubility Information Soluble in water. (1g/L).
Formula Weight 144.21
Percent Purity 99%
Chemical Name or Material n-Butyl butyrate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 203-656-8
RTECSNumber ES8120000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.