Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
n-Butyl Chloride, B&J Brand™, for HPLC, GC and spectrophotometry, ≥99.5%, Honeywell Burdick & Jackson
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
Specifications
Specifications
CAS Max % | 100.0000% |
Density | 0.8862 g/cm3 |
Boiling Point | 78°C |
Flash Point | −9°C |
Loss on Drying | 1 mg/L max. |
Packaging | Glass Bottle |
Quantity | 4 x 4 L |
UN Number | UN1127 |
Beilstein | 1730909 |
Solubility Information | 11% in Water at 20°C |
Show More |
Safety and Handling
ShelfLife | 547 days from date of manufacture |
DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Chlorobutanes |
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction