Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyl sulfide, 99%, pure

Catalog No. p-4509413
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle

CAS: 544-40-1 | C8H18S | 146.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-40-1
Molecular Formula C8H18S
Molecular Weight (g/mol) 146.29
MDL Number MFCD00009468
InChI Key HTIRHQRTDBPHNZ-UHFFFAOYSA-N
Synonym dibutyl sulfide, butyl sulfide, n-butyl sulfide, butylthiobutane, di-n-butyl sulfide, butane, 1,1'-thiobis, butyl monosulfide, dibutyl thioether, thiononane-5, n-dibutyl sulfide
PubChem CID 11002
SMILES CCCCSCCCC

Specifications

Melting Point -80.0°C
Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 188.0°C to 189.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4520 to 1.4540
Linear Formula [CH3(CH2)3]2S
Quantity 100 mL
Beilstein 01,IV,1559
Specific Gravity 0.84
Merck Index 15,1593
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether,soluble in most organic solvents
IUPAC Name 1-butylsulfanylbutane
Formula Weight 146.29
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material n-Butyl sulfide
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 208-870-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.